Learn More
Simport™ Scientific Cryovial™ External Thread with Lip Seal Cryogenic Vials
Shop All Simport Scientific ProductsDescription
- Made of specially formulated polypropylene
- Cap features a long skirt and for one hand aseptic methods
- Fast cap thread design for tightening or removal with a mere 1 1/4 turn and inside thread design will not contribute to contamination
- Lip inside cap ensures leakproof seal even at low temperatures
- Provided with white marking area for sample identification
- Color coded by the use of a CAPINSERT™
- Sterilized by gamma radiation
- Packaged in unique tamperproof, resealable, safety-lock bags
Specifications
Specifications
| Color | Natural |
| Material | Polypropylene |
| Sterility | Sterile |
| Volume (Metric) | 3 mL |
| Thread Style | External |
| Base Style | Self-standing |
| Dnase Free | Yes |
| Autoclavable | Autoclavable |
| Certifications/Compliance | CE; RNase, DNase, Pyrogen and DNA-free |
For in vitro diagnostic (IVD) use only.