Learn More
Simport™ Scientific Cryovial™ External Thread with Lip Seal Cryogenic Vials
Catalog No. p-7112399
Click to view available options
Volume (Metric):
3 mL
4 mL
Designed for storing biological material, human or animal cells, at temperatures as low as -196°C. Cryovial™ External Thread with Lip Seal Cryogenic Vials should be used only in the gas phase of liquid nitrogen.
  • Made of specially formulated polypropylene
  • Cap features a long skirt and for one hand aseptic methods
  • Fast cap thread design for tightening or removal with a mere 1 1/4 turn and inside thread design will not contribute to contamination
  • Lip inside cap ensures leakproof seal even at low temperatures
  • Provided with white marking area for sample identification
  • Color coded by the use of a CAPINSERT™
  • Sterilized by gamma radiation
  • Packaged in unique tamperproof, resealable, safety-lock bags
Specifications
Color Natural
Material Polypropylene
Sterility Sterile
Volume (Metric) 4 mL
Thread Style External
Base Style Self-standing
Dnase Free Yes
Autoclavable Autoclavable
Certifications/Compliance CE; RNase, DNase, Pyrogen and DNA-free
For in vitro diagnostic (IVD) use only.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.