Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Cryptic Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559720
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559720 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559720 Supplier Invitrogen™ Supplier No. PA559720
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: YQREKHNGGR EEVTKVATQK HRQSPLNWTS SHFGEVTGSA EGWGPEEPLP YS Highest antigen sequence identity to the following orthologs: Mouse - 29%, Rat - 33%.

TRUSTED_SUSTAINABILITY

Specifications

Antigen Cryptic
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CFC1
Gene Accession No. P0CG37
Gene Alias AV265756; b2b970Clo; Cfc1; CFC-1; CFC1B; cripto; cripto, FRL-1, cryptic family 1; cryptic; cryptic family 1; Cryptic family protein 1; cryptic protein; CRYPTICFLJ77897; DTGA2; heterotaxy 2 (autosomal dominant); HTX2; HTX2MGC133213
Gene Symbols CFC1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human Cryptic. Recombinant protein control fragment (Product #RP-98817).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55997
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.