Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Cryptic Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA547135
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA547135 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA547135 Supplier Invitrogen™ Supplier No. PA547135
Add to Cart
Add to Cart

Sheep Polyclonal Antibody

In direct ELISAs, approximately 5% cross-reactivity with recombinant mouse Cryptic is observed. Reconstitute at 0.2 mg/mL in sterile PBS.

TRUSTED_SUSTAINABILITY

Specifications

Antigen Cryptic
Applications Flow Cytometry, Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene CFC1
Gene Accession No. P0CG37
Gene Alias AV265756; b2b970Clo; Cfc1; CFC-1; CFC1B; cripto; cripto, FRL-1, cryptic family 1; cryptic; cryptic family 1; Cryptic family protein 1; cryptic protein; CRYPTICFLJ77897; DTGA2; heterotaxy 2 (autosomal dominant); HTX2; HTX2MGC133213
Gene Symbols CFC1
Host Species Sheep
Immunogen Mouse myeloma cell line NS0-derived recombinant human Cryptic Tyr26-Gly169.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55997
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.