Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cryptochrome-1 Rat anti-Mouse, MilliporeSigma™

Catalog No. MABE115225 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABE1152-25 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABE1152-25 Supplier MilliporeSigma Supplier No. MABE115225UL
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Anti-Cryptochrome-1, clone D46-3361 Antibody is a highly specific rat monoclonal antibody, that targets Cryptochrome-1 and has been tested in Western Blotting and Chromatin Immunoprecipitation (ChIP).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cryptochrome-1
Applications ChIP Assay
Classification Monoclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Gene Symbols Cry1
Host Species Rat
Immunogen KLH-conjugated linear peptide corresponding to 20 amino acids from the C-terminal region of murine Cryptochrome-1.
Purification Method Purified by ion-exchange chromatography
Quantity 25 μL
Regulatory Status RUO
Research Discipline Epigenetics and Nuclear Function
Primary or Secondary Primary
Test Specificity Clone D46-3361 is a rat monoclonal antibody that specifically detects Cryptochrome-1 in murine species. it targets an epitope within 20 amino acids from the C-terminal region.
Target Species Mouse
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Isotype IgM
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.