Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Cryptosporidium parvum Oocysts Monoclonal Antibody (E17H)
GREENER_CHOICE

Catalog No. MA17469
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA17469 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA17469 Supplier Invitrogen™ Supplier No. MA17469
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The MA1-7469 antibody reacts with human and bovine fecal oocysts of C. parvum. MA1-7469 has been successfully used in ELISA applications. The MA1-7469 antibody was raised against Whole cryptosporidium Oocysts.

TRUSTED_SUSTAINABILITY

Specifications

Antigen Cryptosporidium parvum Oocysts
Applications ELISA
Classification Monoclonal
Clone E17H
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.2
Gene Alias Cryptosporidiosis
Host Species Mouse
Immunogen Whole Oocysts.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Protozoa
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG3
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.