Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Cryptosporidium parvum Oocysts Monoclonal Antibody (J27E), FITC
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA17471
Description
The MA1-7471 antibody reacts with Cryptosporid. parvum Oocysts in protozoan samples. MA1-7471 has been successfully used in ELISA, Immunohistochemical and Immunofluorescence applications. The MA1-7471 antibody was raised against Whole cryptosporidium Oocysts.

Specifications
Cryptosporidium parvum Oocysts | |
Monoclonal | |
0.1 mg/mL | |
PBS with 10mg/mL BSA and 0.1% sodium azide; pH 7.2 | |
Mouse | |
Protein A | |
RUO | |
Protozoa | |
Antibody | |
IgG1 |
ELISA, Immunohistochemistry | |
J27E | |
FITC | |
Cryptosporidiosis | |
Whole Oocysts. | |
100 μg | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction