Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Cryptosporidium parvum Oocysts Polyclonal Antibody, Biotin
GREENER_CHOICE

Catalog No. PIPA173186
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA173186 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA173186 Supplier Invitrogen™ Supplier No. PA173186
Add to Cart
Add to Cart

Goat Polyclonal Antibody

The PA1-73186 antibody reacts with intact Cryptosporidium parvum oocysts. PA1-73186 has been successfully used in ELISA and Immunofluorescence applications. The PA1-73186 antibody was raised against oocysts purified from bovine feces. Recommended short term storage of less than 6 months at 2-8°C. For extended storage, aliquot and freeze.

TRUSTED_SUSTAINABILITY

Specifications

Antigen Cryptosporidium parvum Oocysts
Applications ELISA, Immunocytochemistry
Classification Polyclonal
Concentration 5 mg/mL
Conjugate Biotin
Formulation PBS with 0.1% sodium azide; pH 7.2
Gene Alias Cryptosporidiosis
Host Species Goat
Immunogen Oocysts purified from bovine feces.
Purification Method IgG fraction
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Protozoa
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.