Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Crystal Violet, 1% w/v in 10% v/v Alcohol, Reagents

Catalog No. 01335727
Click to view available options
Quantity:
20 L

Crystal Violet, 1% w/v in 10% v/v Alcohol, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5, 64-17-5, 64-17-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Gentian Violet, Basic Violet 3
IUPAC Name water
SMILES O

Specifications

Percent Purity 1%
Grade Reagent
Packaging Cubitainer
Specific Gravity 0.984
Color Violet
Quantity 20 L
Physical Form Liquid
Chemical Name or Material Crystal Violet, 1% w/v in 10% v/v Alcohol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.