Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Crystal Violet Indicator, 1% (w/v) in Glacial Acetic Acid, Ricca Chemical
Shop All Ricca Chemical Company Products

Click to view available options
Quantity:
120 mL
500 mL
Packaging:
Amber Glass Bottle
Specifications
Specifications
Concentration | 1% (w/v) |
Percent Purity | 1% W/V |
Grade | Indicator |
Packaging | Amber Glass Bottle |
Solvent or Matrix | Glacial Acetic Acid |
Color | Blue to Purple/Violet |
pH | <2 |
Quantity | 500 mL |
Physical Form | Liquid |
Chemical Name or Material | Crystal Violet (Gentian Violet) Indicator |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction