Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Crystal Violet, pure, high purity biological stain

Catalog No. p-30220
Click to view available options
Quantity:
100 g
500 g

CAS: 548-62-9 | C25H30ClN3 | 407.986 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Crystal Violet
Melting Point 215.0°C
CAS 548-62-9, 90-94-8
Molecular Formula C25H30ClN3
MDL Number MFCD00011750
Beilstein 13, 756
Merck Index 15, 4430
Synonym Basic Violet 3, C.I. 42555, Gentian Violet
Solubility Information Solubility in water: 16g/L (25°C). Other solubilities: soluble in chloroform, 1 g/10 mL alcohol, 1 g/15 mL glycerin, practically insoluble in ether
InChI Key ZXJXZNDDNMQXFV-UHFFFAOYSA-M
SMILES CN(C)C1=CC=C(C=C1)C(=C2C=CC(=[N+](C)C)C=C2)C3=CC=C(C=C3)N(C)C.[Cl-]
Infrared Spectrum Authentic
IUPAC Name [4-[bis[4-(dimethylamino)phenyl]methylidene]cyclohexa-2,5-dien-1-ylidene]-dimethylazanium;chloride
Molecular Weight (g/mol) 407.986
PubChem CID 11057
ChEBI CHEBI:41688
Formula Weight 407.99
Loss on Drying 9% max. (110°C)
Percent Purity ≥96.0% (TiCl3-titration) (on dried substance)
Grade Pure
Packaging Glass bottle
Color Green
Quantity 100 g
Physical Form Powder or Crystals
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.