Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Crystallized Bovine Albumin
Click to view available options
Quantity:
5 g
10 g
100 g
Description
- Source: Bovine blood
- Assay (electrophoresis): ≥99%
- Identity: By IR
- Protein: ≥98%
- pH (7%, 0.15M NaCl): 5.0 - 5.4
- Moisture (KF) when packaged: %le;5.0%
- Sodium (Na): 0 - 5.0mg/g
- Chloride (Cl): 0 - 3.0mg/g
- CAS: 9048-46-8

Specifications
Specifications
pH Range | 5.0 to 5.4 |
Quantity | 5 g |
Source | Bovine blood |
Packaging | Plastic Bottle |
Product Type | Bovine Albumin |
Form | Crystalline |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction