Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CRYZ Mouse anti-Human, Clone: 2A9DD1, Abnova™

Catalog No. 89035544 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-035-544 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-035-544 Supplier Abnova Corporation Supplier No. MAB14146

Mouse Monoclonal Antibody

Mouse monoclonal antibody raised against human CRYZ.

  • High specificity
  • High reactivity

Specifications

Antigen CRYZ
Applications ELISA, Immunocytochemistry, Immunoprecipitation
Classification Monoclonal
Clone 2A9DD1
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against human CRYZ.
Dilution ELISA (2 ug/mL) Immunocytochemistry (2 ug/mL) Immunoprecipitation The optimal working dilution should be determined by the end user.
Formulation In HEPES (0.02% sodium azide).
Gene Accession No. Q08257
Gene Alias DKFZp779E0834/FLJ41475
Gene Symbols CRYZ
Host Species Mouse
Immunogen Human CRYZ.
Purification Method Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1429
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2a κ
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.