Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CRYZ Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA588812
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588812 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588812 Supplier Invitrogen™ Supplier No. PA588812
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MATGQKLMRA VRVFEFGGPE VLKLRSDIAV PIPKDHQVLI KVHACGVNPV ETYIRSGTYS RKPLLPYTPG SDVAGVIEAV GDNASAFKKG DRVFTSSTIS GGYAEYALAA DHTVYKLPEK LDFKQGAAIG IPYFTAYRAL IHSACVKAGE SVLVHGASGG VGLAACQIAR AYGLKILGTA GTEEGQKIVL QNGAHEVFNH REVNYIDKIK KYVGEKGIDI IIEMLANVNL SKDLSLLSHG GRVIVVGSRG TIEINPRDTM AKESSIIGVT LFSSTKEEFQ QYAAALQAGM EIGWLKPVIG SQYPLEKVAE AHENIIHGSG ATGKMILLL; Positive Samples: K-562, Mouse liver, Mouse brain, Mouse kidney, Rat kidney; Cellular Location: Cytoplasm.

Crystallins are separated into two classes: taxon-specific, or enzyme, and ubiquitous. The latter class constitutes the major proteins of vertebrate eye lens and maintains the transparency and refractive index of the lens. The former class is also called phylogenetically-restricted crystallins. This gene encodes a taxon-specific crystallin protein which has NADPH-dependent quinone reductase activity distinct from other known quinone reductases. It lacks alcohol dehydrogenase activity although by similarity it is considered a member of the zinc-containing alcohol dehydrogenase family. Unlike other mammalian species, in humans, lens expression is low. Alternatively spliced transcript variants encoding different isoforms have been found for this gene. One pseudogene is known to exist.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CRYZ
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Cryz
Gene Accession No. P47199, Q08257, Q6AYT0
Gene Alias crystallin zeta; crystallin, zeta; crystallin, zeta (quinone reductase); Cryz; nadph quinone reductase; NADPH:quinone reductase; quinone oxidoreductase; quinone reductase; Sez9; zeta crystallin; Zeta-crystallin
Gene Symbols Cryz
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-329 of human CRYZ (NP_0011235141).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12972, 1429, 362061
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.