Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CRYZL1 Antibody (OTI2F9), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271522F
Description
CRYZL1 Monoclonal specifically detects CRYZL1 in Human, Mouse, Rat, Monkey, Primate samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
CRYZL1 | |
Monoclonal | |
FITC | |
PBS with 0.05% Sodium Azide | |
CRYZL1 | |
Full length recombinant protein of human CRYZL1 (NP_665857) produced in HEK293T cells. | |
0.1 mL | |
9946.0 | |
Store at 4°C in the dark. |
Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
OTI2F9 | |
Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
crystallin, zeta (quinone reductase)-like 1, human quinone oxidoreductase homolog-1104P11QOH-1quinone oxidoreductase-like protein 1, Protein 4P11, Quinone oxidoreductase homolog 1, quinone reductase-like 1, Zeta-crystallin homolog | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat, Monkey, Primate | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction