Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CRYZL1 Antibody (OTI2F9), mFluor Violet 500 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271522MFV500
Description
CRYZL1 Monoclonal antibody specifically detects CRYZL1 in Human, Mouse, Rat, Primate, Monkey samples. It is validated for Flow CytometrySpecifications
CRYZL1 | |
Monoclonal | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Mouse | |
Immunogen affinity purified | |
RUO | |
9946 | |
Store at 4C in the dark. | |
IgG2b |
Flow Cytometry | |
OTI2F9 | |
Flow Cytometry | |
crystallin, zeta (quinone reductase)-like 1, human quinone oxidoreductase homolog-1104P11QOH-1quinone oxidoreductase-like protein 1, Protein 4P11, Quinone oxidoreductase homolog 1, quinone reductase-like 1, Zeta-crystallin homolog | |
Full length human recombinant protein of human CRYZL1(NP_665857) produced in HEK293T cell. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat, Primate, Monkey | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction