Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CRYZL1 Monoclonal Antibody (OTI1D9), TrueMAB™, OriGene
SDP

Mouse Monoclonal Antibody

Supplier:  OriGene TA504608

Encompass_Preferred

 View more versions of this product

Catalog No. 50-167-5260


May include imposed supplier surcharges.
Only null left
Add to Cart

Description

Description

This gene encodes a protein that has sequence similarity to zeta crystallin, also known as quinone oxidoreductase. This zeta crystallin-like protein also contains an NAD(P)H binding site. Alternatively spliced transcript variants have been observed but their full-length nature has not been completely determined.
Specifications

Specifications

CRYZL1
Monoclonal
0.96 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
O95825, Q921W4
CRYZL1
Full length human recombit protein of human CRYZL1 produced in HEK293T cell.
100 μL
Primary
Canine, Human, Mouse, Rat
Antibody
IgG1
Flow Cytometry, Western Blot
OTI1D9
Unconjugated
CRYZL1
4P11, QOH-1
Mouse
Affinity Chromatography
RUO
288256, 478408, 66609, 9946
-20° C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.