Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CRYZL1 Monoclonal Antibody (OTI1H11), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA504768S


Description
CRYZL1 Monoclonal antibody specifically detects CRYZL1 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
CRYZL1 | |
Monoclonal | |
0.9 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
P58340 | |
Mouse | |
Affinity chromatography | |
RUO | |
4291 | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG1 |
Immunohistochemistry (Paraffin), Western Blot | |
OTI1H11 | |
Unconjugated | |
MLF1 | |
2210407J23Rik; 2410006O11Rik; 4P11; crystallin zeta like 1; crystallin, zeta (quinone reductase)-like 1; crystallin, zeta like 1; crystallin, zeta-like 1 isoform a; Cryzl1; dZ63M10.5; protein 4P11; QOH-1; quinone oxidoreductase homolog 1; Quinone oxidoreductase-like protein 1; quinone oxidoreductase-like protein 1; LOW QUALITY PROTEIN: quinone oxidoreductase-like protein 1; quinone reductase-like 1; zeta-crystallin homolog; zgc:92225 | |
Full length human recombinant protein of human MLF1 produced in E.coli. | |
30 μL | |
Primary | |
Human | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction