Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CRYZL1 Monoclonal Antibody (OTI2F9)
GREENER_CHOICE

Catalog No. PIMA525947
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA525947 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA525947 Supplier Invitrogen™ Supplier No. MA525947
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CRYZL1 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow

This gene encodes a protein that has sequence similarity to zeta crystallin, also known as quinone oxidoreductase. This zeta crystallin-like protein also contains an NAD(P)H binding site. Alternatively spliced transcript variants have been observed but their full-length nature has not been completely determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CRYZL1
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone OTI2F9
Concentration 1.08 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene CRYZL1
Gene Accession No. O95825
Gene Alias 2210407J23Rik; 2410006O11Rik; 4P11; crystallin zeta like 1; crystallin, zeta (quinone reductase)-like 1; crystallin, zeta like 1; crystallin, zeta-like 1 isoform a; Cryzl1; dZ63M10.5; protein 4P11; QOH-1; quinone oxidoreductase homolog 1; Quinone oxidoreductase-like protein 1; quinone oxidoreductase-like protein 1; LOW QUALITY PROTEIN: quinone oxidoreductase-like protein 1; quinone reductase-like 1; zeta-crystallin homolog; zgc:92225
Gene Symbols CRYZL1
Host Species Mouse
Immunogen Full length human recombinant protein of CRYZL1 produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9946
Target Species Human, Monkey
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.