Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ CS 2100
GREENER_CHOICESDP

Catalog No. 454350 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45-435-0 50 mg
45-431-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 45-435-0 Supplier Tocris Bioscience™ Supplier No. 4543/50
Add to Cart
Add to Cart
This item is not returnable. View return policy

Selective S1P1 agonist

Sphingosine-1-phosphate receptor 1 (S1P1) agonist (EC50 = 4.0 nM). Exhibits 5000-fold selectivity for human S1P1 over S1P3. Displays efficacy in a rat adjuvant-induced arthritis model.

Chemical Identifiers

CAS 913827-99-3
Molecular Formula C25H23N3O4S

Specifications

Quantity 50 mg
Formula Weight 461.53
Purity >97%
Molecular Weight (g/mol) 461.536
Chemical Name or Material CS 2100
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.