Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CSAD Antibody, mFluor Violet 610 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297312MFV610
Description
CSAD Polyclonal antibody specifically detects CSAD in Human samples. It is validated for Flow CytometrySpecifications
CSAD | |
Polyclonal | |
Flow Cytometry | |
CSDMGC119355, cysteine sulfinic acid decarboxylase, Cysteine-sulfinate decarboxylase, EC 4.1.1, EC 4.1.1.29, FLJ44987, FLJ45500, MGC119354, MGC119357, PCAP, P-selectin cytoplasmic tail-associated protein, Sulfinoalanine decarboxylase | |
Produced in rabbits immunized with E. coli-derived Human CSAD fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
51380 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction