Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CSAD Monoclonal Antibody (2C11)
GREENER_CHOICE

Catalog No. PIMA524559
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA524559 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA524559 Supplier Invitrogen™ Supplier No. MA524559
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: MADSEALPSL AGDPVAVEAL LRAVFGVVVD EAIQKGTSVS QKVCEWKEPE ELKQLLDLEL RSQGESQKQI LERCRAVIRY SVKTGHPRFF NQLFSGLD.

This gene encodes a member of the group 2 decarboxylase family. A similar protein in rodents plays a role in multiple biological processes as the rate-limiting enzyme in taurine biosynthesis, catalyzing the decarboxylation of cysteinesulfinate to hypotaurine. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CSAD
Applications ELISA, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 2C11
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene Csad
Gene Accession No. Q9Y600
Gene Alias Aspartate 1-decarboxylase; CSAD; Csd; cysteine sulfinic acid decarboxylase; cysteine sulfinic acid decarboxylase-related protein; cysteine-sulfinate decarboxylase; PCAP; P-selectin cytoplasmic tail-associated protein; sulfinoalanine decarboxylase
Gene Symbols Csad
Host Species Mouse
Immunogen CSAD (NP_057073, 1 a.a. ~ 98 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Protein A
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51380
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.