Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CSAD Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA597631
Description
CSAD Polyclonal Antibody for Western Blot, ELISA
This gene encodes a member of the group 2 decarboxylase family. A similar protein in rodents plays a role in multiple biological processes as the rate-limiting enzyme in taurine biosynthesis, catalyzing the decarboxylation of cysteinesulfinate to hypotaurine. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene.
Specifications
CSAD | |
Polyclonal | |
Unconjugated | |
Csad | |
Aspartate 1-decarboxylase; CSAD; Csd; cysteine sulfinic acid decarboxylase; cysteine sulfinic acid decarboxylase-related protein; cysteine-sulfinate decarboxylase; PCAP; P-selectin cytoplasmic tail-associated protein; sulfinoalanine decarboxylase | |
Rabbit | |
Protein G | |
RUO | |
246277 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Western Blot | |
3 mg/mL | |
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
Q9DBE0 | |
Csad | |
Recombinant Mouse Cysteine sulfinic acid decarboxylase protein (1-493AA). | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction