Learn More
Invitrogen™ CSAD Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5120240
Description
Positive test controls include: Mouse kidney, Mouse liver, Rat kidney, Rat liver. Immunogen sequence: DVALDTGDKV VQCGRRVDCL KLWLMWKAQG DQGLERRIDQ AFVLARYLVE EMKKREGFEL VMEPEFVNVC FWFVPPSLRG KQESPDYHER LSKVAPVLKE RMVKEGSMMI GYQPHGTRGN FFRVVVANSA LTCADMDFLL NELERLGQDL.
This gene encodes a member of the group 2 decarboxylase family. A similar protein in rodents plays a role in multiple biological processes as the rate-limiting enzyme in taurine biosynthesis, catalyzing the decarboxylation of cysteinesulfinate to hypotaurine. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene.
Specifications
CSAD | |
Polyclonal | |
Unconjugated | |
Csad | |
Aspartate 1-decarboxylase; CSAD; Csd; cysteine sulfinic acid decarboxylase; cysteine sulfinic acid decarboxylase-related protein; cysteine-sulfinate decarboxylase; PCAP; P-selectin cytoplasmic tail-associated protein; sulfinoalanine decarboxylase | |
Rabbit | |
Affinity chromatography | |
RUO | |
246277, 51380, 60356 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
1.65 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
Q64611, Q9DBE0, Q9Y600 | |
Csad | |
Recombinant fusion protein containing a sequence corresponding to amino acids 371-520 of human CSAD (NP_057073.4). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.