Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CSAD Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5145208
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5145208 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5145208 Supplier Invitrogen™ Supplier No. PA5145208
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KECHYSIQKG AAFLGLGTDS VRVVKADERG KMVPEDLERQ IGMAEAEGAV P Antigen sequence identity: Mouse 88%; Rat 90%.

This gene encodes a member of the group 2 decarboxylase family. A similar protein in rodents plays a role in multiple biological processes as the rate-limiting enzyme in taurine biosynthesis, catalyzing the decarboxylation of cysteinesulfinate to hypotaurine. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CSAD
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Csad
Gene Accession No. Q9Y600
Gene Alias Aspartate 1-decarboxylase; CSAD; Csd; cysteine sulfinic acid decarboxylase; cysteine sulfinic acid decarboxylase-related protein; cysteine-sulfinate decarboxylase; PCAP; P-selectin cytoplasmic tail-associated protein; sulfinoalanine decarboxylase
Gene Symbols Csad
Host Species Rabbit
Immunogen Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 188-238. Recombinant protein control fragment (Product #RP-109834).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51380
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Format Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.