Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CSAD Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 228701AP150UL

Description
This gene encodes a member of the group 2 decarboxylase family. A similar protein in rodents plays a role in multiple biological processes as the rate-limiting enzyme in taurine biosynthesis, catalyzing the decarboxylation of cysteinesulfinate to hypotaurine. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene.Specifications
CSAD | |
Polyclonal | |
Unconjugated | |
CSAD | |
CSAD, CSD, PCAP, Sulfinoalanine decarboxylase | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
246277, 51380, 60356 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.37 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q64611, Q9DBE0, Q9Y600 | |
CSAD | |
CSAD Fusion Protein Ag15828 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction