Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CSB Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP303839
Description
CSB Polyclonal specifically detects CSB in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
CSB | |
Polyclonal | |
Immunohistochemistry 1:50-1:200, Immunohistochemistry-Paraffin | |
ARMD5, ATP-dependent helicase ERCC6, CKN2, Cockayne syndrome group B protein, Cockayne syndrome protein CSB, COFS, COFS1, CSBCockayne syndrome B protein, DNA excision repair protein ERCC-6, EC 3.6.1, EC 3.6.4.-, excision repair cross-complementing rodent repair deficiency, complementationgroup 6, RAD26, Rad26 homolog | |
A synthetic peptide corresponding to a sequence within amino acids 700-800 of human CSB (NP_000115.1). LPVFMEQFSVPITMGGYSNASPVQVKTAYKCACVLRDTINPYLLRRMKSDVKMSLSLPDKNEQVLFCRLTDEQHKVYQNFVDSKEVYRILNGEMQIFSGLI | |
100 μL | |
Cancer, DNA Repair, Nucleotide Excision Repair | |
2074 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS with 50% glycerol, pH7.3. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction