Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CSB (ERCC6), Mouse anti-Human, Clone: 1CSB-3H8, Millipore Sigma™

Catalog No. MABE183125 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABE183125 25 μg
MABE1831100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABE183125 Supplier MilliporeSigma Supplier No. MABE183125UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-CSB (ERCC6), clone ICSB-3H8, Cat. No. MABE1831, is a highly specific mouse monoclonal antibody that targets DNA excision repair protein ERCC-6 and has been tested for use in Chromatin Immunoprecipitation (ChIP), Flow Cytometry, Immunocytochemistry, Immunoprecipitation, and Western Blotting.

DNA excision repair protein ERCC-6 (UniProt: Q03468; also known as ATP-dependent helicase ERCC6, Cockayne syndrome protein CSB) is encoded by the ERCC6 (also known as CSB) gene (Gene ID: 2074) in human. CSB is a homodimeric nuclear protein that belongs to the SwItch/ Sucrose nonfermentable family of DNA-dependent ATPases and contains seven characteristic helicase motifs. It is a part of a complex that contains RNA pol I, TFIIH, and XPG, all of which are active in transcription and/or DNA repair. Hence, it is an essential factor involved in transcription-coupled nucleotide excision repair, which allows RNA polymerase II-blocking lesions to be rapidly removed from the transcribed strand of active genes. Upon DNA-binding, it locally modifies DNA conformation by wrapping the DNA around itself, thereby modifying the interface between stalled RNA polymerase II and DNA. It is shown to be essential for transcription-coupled repair complex formation. It recruits the CSA complex (DCX(ERCC8) complex), nucleotide excision repair proteins and EP300 to the at sites of RNA polymerase II-blocking lesions. Mutations in CSB (ERCC6) gene are known to cause age related macular degeneration and Cockayne syndrome B that is characterized by delayed neural development and severe progressive neurologic degeneration and mental retardation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CSB (ERCC6)
Applications ChIP Assay, Flow Cytometry, Immunocytochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 1CSB-3H8
Conjugate Unconjugated
Dilution Immunocytochemistry Analysis: A representative lot detected CSB (ERCC6) in Immunocytochemistry applications (Bradsher, J., et. al. (2002). Mol Cell. 10(4):819-29).Immunoprecipitation Analysis: A representative lot immunoprecipitated CSB (ERCC6) in Immunoprecipitation applications (Bradsher, J., et. al. (2002). Mol Cell. 10(4):819-29).Chromatin Immunoprecipitation (ChIP) Analysis: A representative lot detected CSB (ERCC6) in Chromatin Immunoprecipitation applications (Kristensen, U., et. al. (2013). Proc Natl Acad Sci USA. 110(25):E2261-70; Velez-Cruz, R., et. al. (2012). Proc Natl Acad Sci USA. 110(3):E212-20).Western Blotting Analysis: A representative lot detected CSB (ERCC6) in Western Blotting applications (Bradsher, J., et. al. (2002). Mol Cell. 10(4):819-29).
Formulation Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Gene Alias DNA excision repair protein ERCC-6;ATP-dependent helicase ERCC6;Cockayne syndrome protein CSB
Gene Symbols ERCC6;CSB
Host Species Mouse
Immunogen Ovalbumin-conjugated linear Peptide corresponding to 22 amino acids from the C-terminal region of human DNA excision repair protein ERCC-6.
Purification Method Protein G purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Epigenetics & Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NP_000115.1
Test Specificity Clone 1CSB-3H8 specifically detects DNA excision repair protein ERCC-6 in human cells. It targets an epitope with in 22 amino acids from the C-terminal region.
Target Species Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.