Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CSB Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5111673
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5111673 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5111673 Supplier Invitrogen™ Supplier No. PA5111673
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TLSLHEITDL LETDDSIEAS AIVIQPPENA TAPVSDEESG DEEGGTINNL PGSLLHTAAY LIQDGSDAES DSDDPSYAPK DDSPDEVPST FTVQQPPPSR RRKMTKILCK WKKADLTVQP VAGRVTAPPN DFFTVMRTPT E Highest antigen sequence indentity to the following orthologs: Mouse - 25%, Rat - 25%.

This gene encodes a member of the GTP cyclohydrolase family. The encoded protein is the first and rate-limiting enzyme in tetrahydrobiopterin biosynthesis, catalyzing the conversion of GTP into 7,8-dihydroneopterin triphosphate. BH4 is an essential cofactor required by aromatic amino acid hydroxylases as well as nitric oxide synthases. Mutations in this gene are associated with malignant hyperphenylalaninemia and dopa-responsive dystonia. Several alternatively spliced transcript variants encoding different isoforms have been described; however, not all variants give rise to a functional enzyme.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CSB
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ERCC6
Gene Accession No. Q03468
Gene Alias 4732403I04; ARMD5; ATP-dependent helicase ERCC6; C130058G22Rik; Chimeric CSB-PGBD3 protein; Chimeric ERCC6-PGBD3 protein; CKN2; Cockayne syndrome group B protein; cockayne syndrome protein CSB; COFS; COFS1; CS group B correcting; CSB; DNA excision repair protein ERCC-6; DNA excision repair protein ERCC-6; ERCC6-PGBD3 fusion protein; ERCC excision repair 6, chromatin remodeling factor; ERCC6; ERCC6-PGBD3 fusion protein (isoform 1); excision repair cross-complementation group 6; excision repair cross-complementing rodent repair deficiency, complementation group 6; POF11; RAD26; UVSS1
Gene Symbols ERCC6
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human CSB. Recombinant protein control fragment (Product #RP-108583).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2074
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.