Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

csBAP31 Mouse anti-Human, Unconjugated, Clone: 297-D4, MilliporeSigma™

Catalog No. MABC17112MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABC17112MI 25 μL
MABC17111MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABC17112MI Supplier MilliporeSigma Supplier No. MABC171125UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

csBAP31 Monoclonal antibody specifically detects csBAP31 in Human samples. It is validated for ELISA, Flow Cytometry, Immunoprecipitation, Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen csBAP31
Applications ELISA, Flow Cytometry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 297-D4
Conjugate Unconjugated
Gene Accession No. NP_001132913
Gene Alias Cell surface B-cell receptor-associated protein 31;BCR-associated protein 31;Bap31;6C6-AG tumor-associated antigen;Protein CDM;p28
Gene Symbols BCAP31, BAP31, DXS1357E
Host Species Mouse
Immunogen H9 human embryonic stem cells.
Molecular Weight of Antigen 27.99 kDa calculated.
Purification Method Protein G Purified
Quantity 25 μL
Research Discipline Apoptosis and Cancer
Primary or Secondary Primary
Test Specificity Clone 297-D4 is a mouse monoclonal antibody that detects human Cell surface B-cell receptor-associated protein 31 (csBAP31). It targets an epitope within 10 amino acids from the C-terminal region.
Target Species Human
Content And Storage Stable for 1 year at 2 to 8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.