Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CSDC2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA567430
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA567430 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA567430 Supplier Invitrogen™ Supplier No. PA567430
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MTSESTSPPVV PPLHSPKSPV WP Highest antigen sequence identity to the following orthologs - mouse 96%, rat 100%.

RNA-binding factor which binds specifically to the very 3'-UTR ends of both histone H1 and H3.3 mRNAs, encompassing the polyadenylation signal. Might play a central role in the negative regulation of histone variant synthesis in the developing brain.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CSDC2
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CSDC2
Gene Accession No. Q9Y534
Gene Alias AI415250; AI481750; cold shock domain containing C2; cold shock domain containing C2, RNA binding; cold shock domain-containing protein C2; CSDC2; dJ347H13.2; PIPPIN; RNA-binding protein pippin
Gene Symbols CSDC2
Host Species Rabbit
Immunogen Recombinant Human CSDC2. Recombinant protein control fragment (Product #RP-108092).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27254
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.