Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CSDE1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562571
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562571 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562571 Supplier Invitrogen™ Supplier No. PA562571
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VSFHSHSDHR FLGTVEKEAT FSNPKTTSPN KGKEKEAEDG IIAYDDCGVK LTIAFQAKDV EGSTSPQIGD KVEFSISDKQ RPGQQVATCV RLLGRNSNSK RLLGYVATLK DNFGFIETAN HDKEIFFHYS EFSGDVD Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 99%.

CSDE1 gene ontology annotations related to this gene include regulation of transcription, DNA-templated.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CSDE1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CSDE1
Gene Accession No. O75534
Gene Alias AA960392; BC016898; cold shock domain containing E1; cold shock domain containing E1, RNA binding; cold shock domain containing E1, RNA-binding; cold shock domain-containing protein E1; CSDE1; D1S155E; D3Jfr1; KIAA0885; mKIAA0885; N-ras upstream gene protein; NRAS-related; NRU; Protein UNR; RP5-1000E10.3; UNR; upstream of NRAS
Gene Symbols CSDE1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CSDE1. Recombinant protein control fragment (Product #RP-103118).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7812
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.