Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CSDE1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596480
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596480 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596480 Supplier Invitrogen™ Supplier No. PA596480
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LSAQRNHAIR IKKLPKGTVS FHSHSDHRFL GTVEKEATFS NPKTTSPNKG KEKEAEDGII AYDDCGVKLT IAFQAKDVEG STSPQIGDKV EFSISDKQRP GQQVATCVRL LGRNSNSKRL LGYVATLKDN FGFIETANHD KEIFFHYSEF SGDVDSLELG DMVEYSLSKG KGNKVSAEKV NKTHSVNGIT EEADPTIYSG KVIRPLRSVD PTQTEYQGMI EIVEEGDMKG EV; Positive Samples: U-87MG, LO2, HeLa, MCF7, Mouse brain; Cellular Location: Cytoplasm.

CSDE1 gene ontology annotations related to this gene include regulation of transcription, DNA-templated.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CSDE1
Applications ELISA, Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.74 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene CSDE1
Gene Accession No. O75534, Q91W50
Gene Alias AA960392; BC016898; cold shock domain containing E1; cold shock domain containing E1, RNA binding; cold shock domain containing E1, RNA-binding; cold shock domain-containing protein E1; CSDE1; D1S155E; D3Jfr1; KIAA0885; mKIAA0885; N-ras upstream gene protein; NRAS-related; NRU; Protein UNR; RP5-1000E10.3; UNR; upstream of NRAS
Gene Symbols CSDE1
Host Species Rabbit
Immunogen Recombinant protein (or fragment).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 229663, 7812
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.