Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CSDE1 Recombinant Rabbit Monoclonal Antibody (JE65-67)
GREENER_CHOICE

Catalog No. PIMA544737
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA544737 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA544737 Supplier Invitrogen™ Supplier No. MA544737
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

CSDE1 Recombinant Monoclonal Antibody for Western Blot

CSDE1 gene ontology annotations related to this gene include regulation of transcription, DNA-templated.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CSDE1
Applications Western Blot
Classification Recombinant Monoclonal
Clone JE65-67
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene CSDE1
Gene Accession No. O75534, P18395, Q91W50
Gene Alias AA960392; BC016898; cold shock domain containing E1; cold shock domain containing E1, RNA binding; cold shock domain containing E1, RNA-binding; cold shock domain-containing protein E1; CSDE1; D1S155E; D3Jfr1; KIAA0885; mKIAA0885; N-ras upstream gene protein; NRAS-related; NRU; Protein UNR; RP5-1000E10.3; UNR; upstream of NRAS
Gene Symbols CSDE1
Host Species Rabbit
Immunogen Synthetic peptide within human CSDE1 aa 401-450/798.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 117180, 229663, 7812
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.