Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CSE1L/CAS/Exportin-2 Rabbit anti-Human, Mouse, Clone: 1W4T4, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $458.50
Specifications
Antigen | CSE1L/CAS/Exportin-2 |
---|---|
Clone | 1W4T4 |
Applications | Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
CSE1L/CAS/Exportin-2 Monoclonal antibody specifically detects CSE1L/CAS/Exportin-2 in Human, Mouse samples. It is validated for Western BlotSpecifications
CSE1L/CAS/Exportin-2 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Apoptosis, Cancer, Core ESC Like Genes, Stem Cell Markers | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
1434 | |
IgG | |
Affinity purified |
1W4T4 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse | |
CASMGC117283, Cellular apoptosis susceptibility protein, chromosome segregation 1 (yeast homolog)-like, Chromosome segregation 1-like protein, CSE1, CSE1 chromosome segregation 1-like (yeast), CSE1L, Exp2, exportin-2, Importin-alpha re-exporter, MGC130037, XPO2MGC130036 | |
A synthetic peptide corresponding to a sequence within amino acids 872-971 of human CSE1L/CAS/Exportin-2 (P55060). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title