Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CSF1R Mouse anti-Human, Clone:6B9H2, Abnova™

Catalog No. 89334371 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-334-371 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-334-371 Supplier Abnova Corporation Supplier No. MAB17881
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against partial recombinant human CSF1R.

The protein encoded by this gene is the receptor for colony stimulating factor 1, a cytokine which controls the production, differentiation, and function of macrophages. This receptor mediates most if not all of the biological effects of this cytokine. Ligand binding activates the receptor kinase through a process of oligomerization and transphosphorylation. The encoded protein is a tyrosine kinase transmembrane receptor and member of the CSF1/PDGF receptor family of tyrosine-protein kinases. Mutations in this gene have been associated with a predisposition to myeloid malignancy. The first intron of this gene contains a transcriptionally inactive ribosomal protein L7 processed pseudogene oriented in the opposite direction. [provided by RefSeq]

Specifications

Antigen CSF1R
Applications ELISA, Flow Cytometry, Western Blot
Classification Monoclonal
Clone 6B9H2
Conjugate Unconjugated
Description colony stimulating factor 1 receptor
Dilution ELISA (1:10000)
Flow Cytometry (1:200-1:400)
Western Blot (1:100-1:500)
The optimal working dilution should be determined by the end user.
Formulation In PBS (0.05% sodium azide)
Gene CSF1R
Gene Alias C-FMS, CD115, CSFR, FIM2, FMS
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 344-497 of human CSF1R.
Quantity 100 μg
Gene ID (Entrez) 1436
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.