Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CSF2 Mouse anti-Human, Clone: OTI1A3, lyophilized, TrueMAB™
SDP

Catalog No. 501668598
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-166-8598 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-166-8598 Supplier OriGene Supplier No. CF808006
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO

GM-CSF (Granulocyte-Macrophage colony-stimulating factor) is a 14.6kDa hematopoietic growth factor that exists in glycosylated and non-glycosylated biologically active forms, and stimulates the development of granulocytes, macrophages, early megakaryocytes and eosinophil progenitor cells. The active form of the GM-CSF protein is found extracellularly as a homodimer and the GM-CSF gene has been localized to a cluster of related genes at chromosome region 5q31, which is known to be associated with interstitial deletions in the 5q-syndrome and acute myelogenous leukemia. Other genes in the cluster include those encoding interleukins 4, 5, and 13. The ability of recombit GM-CSF to increase hematopoietic cell recovery has become a focus area in the therapeutic treatment of patients following bone marrow transplantation. Recent studies have investigated GM-CSF in inflammatory and autoimmune diseases such as arthritis, arthritis related interstitial lung disease, nephritis, and psoriasis. In the CNS, GM-CSF depletion or neutralization has been studied in experimental autoimmune encephalomyelitis (EAE) and multiple sclerosis (MS).

Specifications

Antigen CSF2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI1A3
Conjugate Unconjugated
Formulation PBS with 8% trehalose and no preservative; pH 7.3
Gene CSF2
Gene Accession No. P04141
Gene Alias CSF, GMCSF
Gene Symbols CSF2
Host Species Mouse
Immunogen Human recombit protein fragment corresponding to amino acids 18-144 of human CSF2 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1437
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.