Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CSF2 Rat anti-Human, Unconjugated, Clone: BVD2-21C11, Abnova™

Catalog No. 89337393 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-337-393 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-337-393 Supplier Abnova Corporation Supplier No. MAB20896
Add to Cart
Add to Cart

Rat Monoclonal Antibody.

CSF2 Monoclonal antibody specifically detects CSF2 in Human samples. It is validated for WB, IP, IHC-Fr, IF, Flow Cyt, Neutralization.

Specifications

Antigen CSF2
Applications Western Blot, Immunoprecipitation, Immunohistochemistry (Frozen), Immunofluorescence, Flow Cytometry, Neutralization
Classification Monoclonal
Clone BVD2-21C11
Conjugate Unconjugated
Description colony stimulating factor 2 (granulocyte-macrophage)
Gene CSF2
Gene Alias GMCSF, MGC131935, MGC138897
Host Species Rat
Immunogen Recombinant protein corresponding to human CSF2
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 1437
Target Species Human
Content And Storage Store at -20 to -80°C. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.