Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CSF2RA Mouse anti-Human, Clone:4H1, Abnova™

Catalog No. 89333054 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-333-054 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-333-054 Supplier Abnova Corporation Supplier No. MAB16947
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against human CSF2RA.

The protein encoded by this gene is the alpha subunit of the heterodimeric receptor for colony stimulating factor 2, a cytokine which controls the production, differentiation, and function of granulocytes and macrophages. The encoded protein is a member of the cytokine family of receptors. This gene is found in the pseudoautosomal region (PAR) of the X and Y chromosomes. Multiple transcript variants encoding different isoforms have been found for this gene, with some of the isoforms being membrane-bound and others being soluble. [provided by RefSeq]

Specifications

Antigen CSF2RA
Applications CyTOF, Flow Cytometry
Classification Monoclonal
Clone 4H1
Conjugate Unconjugated
Description colony stimulating factor 2 receptor, alpha, low-affinity (granulocyte-macrophage)
Dilution Flow Cytometry
Immunoprecipitation
Immunohistochemistry (Frozen sections)
Mass Cytometry
Western Blot
The optimal working dilution should be determined by the end user.
Formulation In TBS, pH 8.0 (15 mM sodium azide)
Gene CSF2RA
Gene Alias CD116, CDw116, CSF2R, CSF2RAX, CSF2RAY, CSF2RX, CSF2RY, GM-CSF-R-alpha, GMCSFR, GMR, MGC3848, MGC4838
Host Species Mouse
Immunogen CSF2RA-transfected COS cells.
Quantity 100 μg
Gene ID (Entrez) 1438
Test Specificity This antibody recognizes human CD116, the GM-CSF receptor alpha subunit (approxmately 80 kDa) expressed by neutrophils, eosinophils, monocytes and macrophages.
Target Species Human
Content And Storage Store at 4°C. Do not freeze.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.