Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CSF2RB Chimeric Recombinant Rabbit Monoclonal Antibody (BION-1)
GREENER_CHOICE

Catalog No. PIMA548120
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA548120 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA548120 Supplier Invitrogen™ Supplier No. MA548120
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Specificity: Binds to domain 4 of the common beta-chain and recognises the same binding site used by IL-5, GM-CSF and IL-3, or binds to an epitope in close proximity to it. Residues M363, R364, E366 (B-C loop) and R418 (F-G loop) in the common beta-chain are important for BION-1 binding. Preparation: BALB/c mice were immunized intraperitonally with COS cells transfected DQP (Domain 4 of the common beta-chain) expression constructs. The immunizations were repeated 4 times at 2-week intervals. Four weeks after the final immunization, a mouse was boosted with COS transfectants intravenously. Three days later, splenocytes were harvested and fused with NS-1 myeloma cells. Hybridoma supernatants were screened on CHO DQP cells by flow cytometry, with untransfected CHO cells as a control. All antibodies were from single hybridoma clones as selected by a limiting dilution method. MoAbs were purified from ascites fluid or hybridoma supernatant by a protein A sepharose column.

The protein encoded by this gene is the common beta chain of the high affinity receptor for IL-3, IL-5 and CSF. Defects in this gene have been reported to be associated with protein alveolar proteinosis (PAP).
TRUSTED_SUSTAINABILITY

Specifications

Antigen CSF2RB Chimeric
Applications Flow Cytometry, Functional Assay, Inhibition Assays, Immunohistochemistry, Immunoprecipitation, Immunocytochemistry
Classification Recombinant Monoclonal
Clone BION-1
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% ProClin 300
Gene Csf2rb
Gene Accession No. P32927
Gene Alias Ai2ca; AI848964; AIC2A; Aic2b; Bc; beta c; beta common cytokine receptor; BetaIl3; Bil3; CD antigen CD131; CD131; CD131 antigen; CDw131; colony stimulating factor 2 receptor beta common subunit; colony stimulating factor 2 receptor, beta 1, low-affinity (granulocyte-macrophage); colony stimulating factor 2 receptor, beta 2, low-affinity (granulocyte-macrophage); colony stimulating factor 2 receptor, beta, low-affinity (granulocyte-macrophage); colony-stimulating factor 2 receptor subunit beta-2; colony-stimulating factor-2 receptor, beta, low-affinity; common beta chain; CSF2RB; Csf2rb1; Csf2rb2; Csfgmrb; cytokine receptor common subunit beta; GM-CSF/IL-3/IL-5 receptor common beta subunit; GM-CSF/IL-3/IL-5 receptor common beta-chain; IL-3 RB; IL-3 Receptor beta; IL-3 receptor class 2 subunit beta; Il3r; IL3R beta; IL-3R class 2 subunit beta; IL3RB; Il3rb1; Il3rb2; IL5RB; interleukin 3 common beta chain; interleukin 3 receptor/granulocyte-macrophage colony stimulating factor 3 receptor, beta (high affinity); interleukin-3 receptor class 2 subunit beta; Interleukin-3 receptor class II beta chain; SMDP5
Gene Symbols Csf2rb
Host Species Rabbit
Immunogen BALB/c mice were immunized intraperitonally with COS cells transfected DQP (Domain 4 of the common beta-chain) expression constructs.
Purification Method Protein A
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1439
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.