Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CSF2RB Monoclonal Antibody (OTI7G6), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA807873
Description
CSF2RB is the common beta chain of the high affinity receptor for IL-3, IL-5 and CSF. Defects in this gene have been reported to be associated with protein alveolar proteinosis (PAP). Diseases associated with CSF2RB include Surfactant Metabolism Dysfunction, Pulmonary, 5 and Hereditary Pulmonary Alveolar Proteinosis.Specifications
| CSF2RB | |
| Monoclonal | |
| Unconjugated | |
| CSF2RB | |
| CD131, CDw131, IL3RB, IL5RB, SMDP5, betaGMR | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 1439 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI7G6 | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P32927 | |
| CSF2RB | |
| Full length human recombit protein of human CSF2RB produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction