Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CSF3R Monoclonal Antibody (8F8B12)
GREENER_CHOICE

Catalog No. FERMA548432
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FERMA548432 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FERMA548432 Supplier Invitrogen™ Supplier No. MA548432
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CSF3R Monoclonal Antibody for ELISA, Flow, ICC/IF, IHC (P), Western Blot

CSF3R is the receptor for colony stimulating factor 3, a cytokine that controls the production, differentiation, and function of granulocytes. The encoded protein, which is a member of the family of cytokine receptors, may also function in some cell surface adhesion or recognition processes. Four transcript variants encoding four different isoforms have been found for this gene, with three of the isoforms being membrane-bound and the other being secreted and soluble.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CSF3R
Applications ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 8F8B12
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene CSF3R
Gene Accession No. Q99062
Gene Alias CD114; CD114 antigen; colony stimulating factor 3 receptor; colony stimulating factor 3 receptor (granulocyte); Csf3r; Csfgr; G-CSF receptor; GCSFR; G-CSFR; G-CSF-R; Granulocyte colony-stimulating factor receptor; SCN7
Gene Symbols CSF3R
Host Species Mouse
Immunogen Purified recombinant fragment of human CD114 (AA: extra 25-187) expressed in E. Coli.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1441
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.