Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CSF3R Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5119914
This item is not returnable.
View return policy
Description
Positive Control: HL-60 cell lysate, THP-1 cell lysate, SH-SY5Y cell lysate, Hela. Subcellular Location: Secreted, Cell membrane.
CSF3R is the receptor for colony stimulating factor 3, a cytokine that controls the production, differentiation, and function of granulocytes. The encoded protein, which is a member of the family of cytokine receptors, may also function in some cell surface adhesion or recognition processes. Four transcript variants encoding four different isoforms have been found for this gene, with three of the isoforms being membrane-bound and the other being secreted and soluble.
Specifications
CSF3R | |
Polyclonal | |
Unconjugated | |
CSF3R | |
CD114; CD114 antigen; colony stimulating factor 3 receptor; colony stimulating factor 3 receptor (granulocyte); Csf3r; Csfgr; G-CSF receptor; GCSFR; G-CSFR; G-CSF-R; Granulocyte colony-stimulating factor receptor; SCN7 | |
Rabbit | |
Affinity chromatography | |
RUO | |
1441 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Flow Cytometry, Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q99062 | |
CSF3R | |
Synthetic peptide within human GCSF Receptor aa 787-836. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction