Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CSF3R Polyclonal Antibody

Goat Polyclonal Antibody
Supplier: Invitrogen™ PA5142945
This item is not returnable.
View return policy
Description
Additional Information: This antibody is expected to recognize all reported isoforms (NP_000751.1; NP_724781.1; NP_758519.1). This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:128,000.
CSF3R is the receptor for colony stimulating factor 3, a cytokine that controls the production, differentiation, and function of granulocytes. The encoded protein, which is a member of the family of cytokine receptors, may also function in some cell surface adhesion or recognition processes. Four transcript variants encoding four different isoforms have been found for this gene, with three of the isoforms being membrane-bound and the other being secreted and soluble.
Specifications
CSF3R | |
Polyclonal | |
Unconjugated | |
CSF3R | |
CD114; CD114 antigen; colony stimulating factor 3 receptor; colony stimulating factor 3 receptor (granulocyte); Csf3r; Csfgr; G-CSF receptor; GCSFR; G-CSFR; G-CSF-R; Granulocyte colony-stimulating factor receptor; SCN7 | |
Goat | |
Ammonium Sulfate Precipitation | |
RUO | |
1441 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
Q99062 | |
CSF3R | |
Peptide with sequence C-PPELGKSPLTHYT. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction