Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CSGALNACT1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501982486
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-2486 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-2486 Supplier Bioss Supplier No. BS12938R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CSGALNACT1 transfers 1,4-N-acetylgalactosamine (GalNAc) from UDP-GalNAc to the non-reducing end of glucuronic acid (GlcUA). CSGALNACT1 required for addition of the first GalNAc to the core tetrasaccharide linker and for elongation of chondroitin chains. CSGALNACT1 is important role in chondroitin chain biosynthesis in cartilage.

Specifications

Antigen CSGALNACT1
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene CSGALNACT1
Gene Accession No. Q8TDX6
Gene Alias 4732435N03Rik; beta1,4 N-acetylgalactosaminyltransferase; beta4GalNAcT; Beta4GalNAcT-1; Chgn; chondroitin beta1,4 N-acetylgalactosaminyltransferase; Chondroitin beta-1,4-N-acetylgalactosaminyltransferase 1; Chondroitin sulfate N-acetylgalactosaminyltransferase 1; Csgalnact1; CsGalNAcT-1; GALNACT1; glucuronylgalactosylproteoglycan 4-beta-N- acetylgalactosaminyltransferase; UNQ656/PRO1287
Gene Symbols CSGALNACT1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human CSGALNACT1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55790
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.