Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CSGALNACT2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA543328
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA543328 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA543328 Supplier Invitrogen™ Supplier No. PA543328
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: LYNPAIVYAN QEVPPPVEQQ LVHKKDSGFW RDFGFGMTCQ YRSDFLTIGG Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 77%.

This gene encodes a member of the chondroitin N-acetylgalactosaminyltransferase family. The encoded protein is involved in elongation during chondroitin sulfate synthesis. A related pseudogene has been identified on chromosome X.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CSGALNACT2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene Csgalnact2
Gene Accession No. Q8N6G5
Gene Alias 4632415D10Rik; beta 4 GalNAcT-2; beta4GalNAcT; Beta4GalNAcT-2; CHGN2; ChGn-2; chondroitin beta1,4 N-acetylgalactosaminyltransferase 2; Chondroitin beta-1,4-N-acetylgalactosaminyltransferase 2; chondroitin sulfate GalNAcT-2; chondroitin sulfate N-acetylgalactosaminyltransferase 2; CSGALNACT2; Galnact2; galNAcT-2; glucuronylgalactosylproteoglycan 4-beta-N-acetylgalactosaminyltransferase 2; PRO0082; RGD1563660
Gene Symbols Csgalnact2
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human CSGALNACT2 (aa 398-447).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55454
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.