Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CSGALNACT2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556773
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556773 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556773 Supplier Invitrogen™ Supplier No. PA556773
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GKEGLSKVKS ILESVTSESN FHNYTLVSLN EEFNRGRGLN VGARAWDKGE VLM Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 94%.

This gene encodes a member of the chondroitin N-acetylgalactosaminyltransferase family. The encoded protein is involved in elongation during chondroitin sulfate synthesis. A related pseudogene has been identified on chromosome X.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CSGALNACT2
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Csgalnact2
Gene Accession No. Q8N6G5
Gene Alias 4632415D10Rik; beta 4 GalNAcT-2; beta4GalNAcT; Beta4GalNAcT-2; CHGN2; ChGn-2; chondroitin beta1,4 N-acetylgalactosaminyltransferase 2; Chondroitin beta-1,4-N-acetylgalactosaminyltransferase 2; chondroitin sulfate GalNAcT-2; chondroitin sulfate N-acetylgalactosaminyltransferase 2; CSGALNACT2; Galnact2; galNAcT-2; glucuronylgalactosylproteoglycan 4-beta-N-acetylgalactosaminyltransferase 2; PRO0082; RGD1563660
Gene Symbols Csgalnact2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CSGALNACT2. Recombinant protein control fragment (Product #RP-95690).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55454
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.