Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CSGlcAT Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA543234
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA543234 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA543234 Supplier Invitrogen™ Supplier No. PA543234
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: SLLRVSWIQG EGEDPCVEAV GERGGPQNPD SRARLDQSDE DFKPRIVPYY Sequence homology: Dog: 79%; Horse: 93%; Human: 100%; Pig: 79%; Rabbit: 79%.

CSGlcA-T belongs to the chondroitin N-acetylgalactosaminyltransferase family. It transfers glucuronic acid (GlcUA) from UDP-GlcUA to N-acetylgalactosamine residues on the non-reducing end of the elongating chondroitin polymer. CSGlcA-T has no N-acetylgalactosaminyltransferase activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CSGlcAT
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene CHPF2
Gene Accession No. Q9P2E5
Gene Alias Chondroitin glucuronyltransferase; Chondroitin polymerizing factor 2; chondroitin sulfate glucuronyltransferase; Chondroitin synthase 3; chondroitin synthase-3; CHPF2; chPF-2; CHSY3; ChSy-3; CSGlcAT; CSGlcA-T; KIAA1402; N-acetylgalactosaminyl-proteoglycan 3-beta-glucuronosyltransferase; UNQ299/PRO339
Gene Symbols CHPF2
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human CSGlcA-T (aa 27-76).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54480
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.