Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CSGlcAT Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA554298
Description
Immunogen sequence: PCVEAVGERG GPQNPDSRAR LDQSDEDFKP RIVPYYRDPN KPYKKVLRTR YIQTELGSRE RLLVAVLTSR ATLST Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 91%.
CSGlcA-T belongs to the chondroitin N-acetylgalactosaminyltransferase family. It transfers glucuronic acid (GlcUA) from UDP-GlcUA to N-acetylgalactosamine residues on the non-reducing end of the elongating chondroitin polymer. CSGlcA-T has no N-acetylgalactosaminyltransferase activity.
Specifications
CSGlcAT | |
Polyclonal | |
Unconjugated | |
CHPF2 | |
Chondroitin glucuronyltransferase; Chondroitin polymerizing factor 2; chondroitin sulfate glucuronyltransferase; Chondroitin synthase 3; chondroitin synthase-3; CHPF2; chPF-2; CHSY3; ChSy-3; CSGlcAT; CSGlcA-T; KIAA1402; N-acetylgalactosaminyl-proteoglycan 3-beta-glucuronosyltransferase; UNQ299/PRO339 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
54480 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9P2E5 | |
CHPF2 | |
Recombinant protein corresponding to Human CSGlcAT. Recombinant protein control fragment (Product #RP-92616). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction