Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CSGLCAT Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5101521
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5101521 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5101521 Supplier Invitrogen™ Supplier No. PA5101521
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total CSGLCAT.

CSGlcA-T belongs to the chondroitin N-acetylgalactosaminyltransferase family. It transfers glucuronic acid (GlcUA) from UDP-GlcUA to N-acetylgalactosamine residues on the non-reducing end of the elongating chondroitin polymer. CSGlcA-T has no N-acetylgalactosaminyltransferase activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CSGLCAT
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene CHPF2
Gene Accession No. Q9P2E5
Gene Alias Chondroitin glucuronyltransferase; Chondroitin polymerizing factor 2; chondroitin sulfate glucuronyltransferase; Chondroitin synthase 3; chondroitin synthase-3; CHPF2; chPF-2; CHSY3; ChSy-3; CSGlcAT; CSGlcA-T; KIAA1402; N-acetylgalactosaminyl-proteoglycan 3-beta-glucuronosyltransferase; UNQ299/PRO339
Gene Symbols CHPF2
Host Species Rabbit
Immunogen A synthesized peptide derived from human CHPF2(Accession Q9P2E5), corresponding to amino acid residues G36-V86.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54480
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.